Order Entry
Italy
ContactUsLinkComponent
 
 
 
4-Acetamidobenzenesulphonyl chloride ≥98.0% (by HPLC, titration analysis)
  TCIAA0074-25G
 :  TCI
 :  
undefined
 
 
4-Acetamidobenzenesulphonyl chloride ≥98.0% (by HPLC, titration analysis)
  TCIAA0074-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

 

 :
 :233.68 g/mol
 :14,00103
 :416 °C (1013 hPa)
 :146 °C
 :204-485-1
 :3261
 :8,III
 :GHS05
 :GHS07
 :121-60-8
 :C₈H₈ClNO₃S
 :MFCD00007442