
4,4'-Diaminobenzophenone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:611-98-3
:
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00038138
:212.25 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:245.0 to 249.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together