Order Entry
Italy
ContactUsLinkComponent
3,4-Dimethoxybenzonitrile ≥98.0% (by GC)
Toxic
3,4-Dimethoxybenzonitrile ≥98.0% (by GC)
Catalog # TCIAD2870-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
3,4-Dimethoxybenzonitrile ≥98.0% (by GC)
Catalog # TCIAD2870-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    67.0 to 71.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :163.18 g/mol
Boiling Pt:131 - 133 °C (0,3 -rr)
Melting Pt:71…72 °C
EINECS:217-969-2
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:2024-83-1
Formula:C₉H₉NO₂
MDL Number:MFCD00001802