Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
Frequently Bought Together