
2-Chloro-1-methylpyridinium iodide ≥98.0% (by titrimetric analysis)
TCIAC0903-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
:
:255.49 g/mol
:14,06301
:207 °C
:238-288-7
:GHS07
:14338-32-0
:C₆H₇ClIN
:MFCD00011984