Order Entry
Italy
ContactUsLinkComponent
α-Chloro-4-(trifluoromethoxy)toluene ≥98.0% (by GC)
Corrosive
α-Chloro-4-(trifluoromethoxy)toluene ≥98.0% (by GC)
Catalog # TCIAT2453-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
α-Chloro-4-(trifluoromethoxy)toluene ≥98.0% (by GC)
Catalog # TCIAT2453-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :210.58
Boiling Pt:90°C
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
 :GHS05
CAS Number:65796-00-1
Formula:C₈H₆ClF₃O
MDL Number:MFCD00052326