
3-Acetoxybenzaldehyde ≥98.0% (by GC)
TCIAA2467-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
:
:164.16 g/mol
:110°C
:251-890-4
:GHS07
:34231-78-2
:C₉H₈O₃
:MFCD00016603