
2,5-Dibromo-3-picoline ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3430-18-0
:
Formula:C₆H₅Br₂N
MDL Number:MFCD02093085
Boiling Pt:114°C
ADR:6.1,III
:250.92 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:44.0 to 47.0 deg-C
Specification Test Results
Frequently Bought Together