Order Entry
Italy
ContactUsLinkComponent
Cyclopropyltriphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Irritant
Cyclopropyltriphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAC1378-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Cyclopropyltriphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAC1378-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    184.0 to 187.0 deg-C

Specification Test Results

About this item

 :
 :383.27 g/mol
Melting Pt:189…190 °C
EINECS:237-970-1
 :GHS07
CAS Number:14114-05-7
Formula:C₂₁H₂₀BrP
MDL Number:MFCD00011872