
1,2-Dibromotetrachloroethane ≥97.0% (by GC)
Catalog # TCIAD1987-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:325.64 g/mol
Boiling Pt:222 °C (1013 hPa)
Melting Pt:83 °C
Density:2,713 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-136-7
:GHS07
CAS Number:630-25-1
Formula:BrCl₂CCBrCl₂
MDL Number:MFCD00000780