Order Entry
Italy
ContactUsLinkComponent
 
 
2,3-Dibromobutane ≥99.0% (by GC)
  TCIAD0177-25G
 :  TCI
 :  
undefined
 
2,3-Dibromobutane ≥99.0% (by GC)
  TCIAD0177-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :215.92 g/mol
 :157°C
 :–24 °C
 :1,83 g/cm³ (0 °C)
 :65°C
 :226-476-1
 :GHS07
 :5408-86-6
 :H₃C(BrCH)₂CH₃
 :MFCD00000146