Order Entry
Italy
ContactUsLinkComponent
1,9-Dibromononane ≥97.0% (by GC)
Irritant
1,9-Dibromononane ≥97.0% (by GC)
Catalog # TCIAD1746-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,9-Dibromononane ≥97.0% (by GC)
Catalog # TCIAD1746-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :286.05 g/mol
Boiling Pt:288°C
Melting Pt:-2°C
Melting Pt:–2°C
Density:1,41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:224-913-0
 :GHS07
CAS Number:4549-33-1
Formula:BrH₂C(CH₂)₇CH₂Br
MDL Number:MFCD00000279