
1,7-Dibromoheptane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4549-31-9
:
Formula:BrH₂C(CH₂)₅CH₂Br
MDL Number:MFCD00000274
:258 g/mol
Boiling Pt:263°C
Melting Pt:–41.7 °C
Density:1,5451 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:224-910-4
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together