
1,6-Dibromohexane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:629-03-8
:
Formula:BrH₂C(CH₂)₄CH₂Br
MDL Number:MFCD00000272
:243.97 g/mol
Boiling Pt:246°C
Melting Pt:–2°C
Density:1,61 g/cm³ (20 °C)
Flash Pt:152°C
Storage Temperature:Ambient
EINECS:211-067-2
ADR:9,III
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together