Order Entry
Italy
ContactUsLinkComponent
 
 
1,4-Dibromopentane ≥95.0% (by GC)
 :  TCI
 
undefined
 
1,4-Dibromopentane ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together