
1,3-Dibromobutane ≥98.0% (by GC)
TCIAD0175-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
:
:215.92 g/mol
:175°C
:1,789 g/cm³ (20 °C)
:203-520-8
:GHS07
:Ambient
:107-80-2
:BrH₂CCH₂BrCHCH₃
:MFCD00000152