

1,2-Dibromopropane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:78-75-1
:
Formula:BrH₂CBrCHCH₃
MDL Number:MFCD00000148
:201.89 g/mol
Merck Index:14,07853
Boiling Pt:141°C
Melting Pt:–55 °C
Density:1,932 g/cm³ (20 °C)
Flash Pt:50°C
Storage Temperature:Ambient
EINECS:201-139-1
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together