Order Entry
Italy
ContactUsLinkComponent
 
Irritant
1,2-Dibromobutane ≥98.0% (by GC)
Catalog # TCIAD0174-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,2-Dibromobutane ≥98.0% (by GC)
Catalog # TCIAD0174-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :215.92 g/mol
Merck Index:14,01566
Boiling Pt:166°C
Melting Pt:–64 °C
Density:0,8262 g/cm³ (0 °C)
EINECS:208-581-4
 :GHS07
CAS Number:533-98-2
Formula:BrH₂CBrCHCH₂CH₃
MDL Number:MFCD00000157