Order Entry
Italy
ContactUsLinkComponent
1,2-Dihydronaphthalene ≥98.0% (by GC)
1,2-Dihydronaphthalene ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
1,2-Dihydronaphthalene ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together