
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98.0% (by GC)
: TCI
:355-80-6
:
:C₅H₄F₈O
:MFCD00039631
:232.07 g/mol
:14,06746
:140°C
:< –50 °C
:1,663 g/cm³ (20 °C)
:206-593-4
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together