
Xylene cyanole FF
Supplier: TCI
Certificates
About this item
CAS Number:2650-17-1
:
Formula:C₂₅H₂₇N₂NaO₆S₂
MDL Number:MFCD00040657
:538.62 g/mol
Melting Pt:295 °C
EINECS:220-167-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Green to Dark green powder to crystal
- Lambda max.:613.0 to 616.0 nm (H2O)
- Absorbance (E1%1cm):min. 950 (H2O, 613.0 to 616.0 nm)
Specification Test Results
Frequently Bought Together