
2,4'-Difluorobenzophenone ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:342-25-6
:
Formula:C₁₃H₈F₂O
MDL Number:MFCD00000319
:218.2 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:22…24 °C
Density:1,24 g/cm³ (20 °C)
EINECS:206-441-7
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 96.0 %
- Freezing point:22.0 to 28.0 deg-C
Specification Test Results
Frequently Bought Together