
2,2',4,4'-Tetrahydroxybenzophenone ≥98.0% (by HPLC)
TCIAT0118-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Orange to Green to Amber powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:197.0 to 202.0 deg-C
- Solubility in Methanol:almost transparency
:
:246.22 g/mol
:100 - 102 °C (40 -rr)
:196…198 °C
:1,498 g/cm³ (20 °C)
:205-028-9
:GHS07
:131-55-5
:C₁₃H₁₀O₅
:MFCD00002278