
2-Bromomethyl-1,4-benzodioxane ≥98.0% (by GC, titration analysis)
Catalog # TCIAB2349-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:229.07 g/mol
Boiling Pt:63 - 64 °C (0,005 -rr)
EINECS:218-503-0
:GHS07
CAS Number:2164-34-3
Formula:C₉H₉BrO₂
MDL Number:MFCD00044831