3-Phenoxy-1,2-propandiol ≥95.0% (by GC)
: TCI
:538-43-2
:
:C₆H₅OCH₂CH(OH)CH₂OH
:MFCD00004717
:168.19 g/mol
:14,07290
:202°C
:70 °C
:1,1477 g/cm³ (20 °C)
:208-692-8
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:54.0 to 58.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together