Order Entry
Italy
ContactUsLinkComponent
4-Methoxy-1-naphthaldehyde ≥97.0% (by GC)
4-Methoxy-1-naphthaldehyde ≥97.0% (by GC)
  TCIAM1049-5G
 :  TCI
 :  
undefined
4-Methoxy-1-naphthaldehyde ≥97.0% (by GC)
  TCIAM1049-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

 

 

  • Appearance:
    Light orange to Yellow to Green powder to lump
  • Purity (GC):
    min. 97.0 %

 

 

 : 4-Methoxynaphthalene-1-carbaldehyde
 :
 :186.21 g/mol
 :240-109-2
 :15971-29-6
 :C₁₂H₁₀O₂
 :MFCD00004006
 :321
 :36