Order Entry
Italy
ContactUsLinkComponent
 
 
3,6-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
3,6-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

 

Frequently Bought Together