

EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) ≥98.0% (by GC, titration analysis)
: TCI
:1892-57-5
:
:C₈H₁₇N₃
:MFCD00044916
:155.24 g/mol
:45°C
:114…115 °C
:0,885 g/cm³ (20 °C)
:Freezer
:217-579-2
:2735
:8,III
:GHS05
:GHS07
- Appearance:Colorless to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together