Order Entry
Italy
ContactUsLinkComponent
1,10-Phenanthroline hydrochloride monohydrate ≥99.0% (by titrimetric analysis)
Irritant
Environment
1,10-Phenanthroline hydrochloride monohydrate ≥99.0% (by titrimetric analysis)
Catalog # TCIAP0081-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Environment
1,10-Phenanthroline hydrochloride monohydrate ≥99.0% (by titrimetric analysis)
Catalog # TCIAP0081-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency
  • Suitability for iron-analysis:
    to pass test

Specification Test Results

About this item

 :
 :234.68 g/mol
Melting Pt:224…226 °C
Density:1,385 g/cm³ (-100.15 °C)
Storage Temperature:Ambient
EINECS:223-325-1
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:18851-33-7
 :3829-86-5
Formula:C₁₂H₉ClN₂·1H₂O
MDL Number:MFCD00150061