
(±)-trans-1,2-Diaminocyclohexane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1121-22-8
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00063747
:114.19 g/mol
Boiling Pt:80°C
Melting Pt:14.8 °C
Density:0,951 g/cm³ (20 °C)
Flash Pt:68°C
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together