Order Entry
Italy
ContactUsLinkComponent
2-Acetyl-5-methylthiophene ≥96.0% (by GC)
Irritant
2-Acetyl-5-methylthiophene ≥96.0% (by GC)
Catalog # TCIAA1181-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Acetyl-5-methylthiophene ≥96.0% (by GC)
Catalog # TCIAA1181-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :140.21 g/mol
Boiling Pt:65 - 67 °C (1 -rr)
Melting Pt:24…28 °C
Density:1,119 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:237-181-2
 :GHS07
CAS Number:13679-74-8
Formula:C₇H₈OS
MDL Number:MFCD00014529