Order Entry
Italy
ContactUsLinkComponent
4-Bromo-2,6-di-tert-butylphenol ≥98.0% (by GC)
Irritant
4-Bromo-2,6-di-tert-butylphenol ≥98.0% (by GC)
Catalog # TCIAB1758-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Bromo-2,6-di-tert-butylphenol ≥98.0% (by GC)
Catalog # TCIAB1758-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    82.0 to 85.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :285.22 g/mol
Boiling Pt:128°C
Melting Pt:83 °C
EINECS:214-521-8
 :GHS07
CAS Number:1139-52-2
Formula:C₁₄H₂₁BrO
MDL Number:MFCD00051598