
3,5-Di-tert-butylpyrocatechol ≥98.0% (by GC)
Catalog # TCIAD2432-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:99.0 to 102.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
Boiling Pt:145°C
Melting Pt:96…97 °C
:222.33 g/mol
EINECS:213-816-9
:GHS07
CAS Number:1020-31-1
Formula:C₁₄H₂₂O₂
MDL Number:MFCD00008819