Cholesterol ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:57-88-5
:
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646
:386.66 g/mol
Merck Index:14,02201
Boiling Pt:360 °C (1013 hPa)
Melting Pt:147…150 °C
Density:1,07 g/cm³ (20 °C)
:Stabilised with α-tocopherol
Storage Temperature:Refrigerator
EINECS:200-353-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:148.0 to 150.0 deg-C
- Specific rotation [a]25/D:-34.0 to -39.5 deg (C=2, Dioxane)
- Solubility in hot EtOH:very faint turbidity
Specification Test Results
Frequently Bought Together