Dicyclohexyl sulphide ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2550-40-5
:
Formula:C₁₂H₂₂S₂
MDL Number:MFCD00013759
:230.44 g/mol
Boiling Pt:110°C
Density:1,0457 g/cm³ (20 °C)
EINECS:219-851-6
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 90.0 %
Specification Test Results
Frequently Bought Together