Order Entry
Italy
ContactUsLinkComponent
2,6-Dinitro-p-cresol stabilised
2,6-Dinitro-p-cresol stabilised
Catalog # EHERC12784000
Supplier:  EHRENSTORFER
CAS Number:  
2,6-Dinitro-p-cresol stabilised
Catalog # EHERC12784000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :198.14 g/mol
Melting Pt:77…79 °C
 :Moistened with water
EINECS:210-203-8
CAS Number:609-93-8
Formula:C₇H₆N₂O₅
MDL Number:MFCD00007121