

1,3-Diaminopentane ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:589-37-7
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00134550
:102.18 g/mol
Boiling Pt:164°C
Flash Pt:59°C
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together