Triethylene glycol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol
CAS Number:112-27-6
:
Formula:C₆H₁₄O₄
MDL Number:MFCD00002880
Boiling Pt:276°C
Molecular Weight:150.17 g/mol
Melting Pt:-7
Flash Pt:174°C
Merck Index:14,09670
EINECS:203-953-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together