Order Entry
Italy
ContactUsLinkComponent
Tetramethylammonium hexafluorophosphate
Irritant
Tetramethylammonium hexafluorophosphate
Catalog # TCIAT1296-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Tetramethylammonium hexafluorophosphate
Catalog # TCIAT1296-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 99.0 %
  • Solubility in hot Water:
    almost transparency

Specification Test Results

About this item

 :
 :219.11 g/mol
Melting Pt:300 °C
Density:1,617 g/cm³ (25 °C)
EINECS:209-194-3
ADR:8,III
 :GHS07
CAS Number:558-32-7
Formula:C₄H₁₂F₆NP
MDL Number:MFCD00012012