Propyl propionate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-36-5
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009373
:116.16 g/mol
Merck Index:14,07867
Boiling Pt:122°C
Density:0,88 g/cm³ (20 °C)
Flash Pt:27°C
EINECS:203-389-7
UN:3272
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together