
Methacrylamide ≥98.0% (by GC, titration analysis)
: TCI
:79-39-0
:
:C₄H₇NO
:MFCD00008018
:85.11 g/mol
:215 °C (1013 hPa)
:106…109 °C
:1,053 g/cm³ (20 °C)
:103 °C
:201-202-3
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Bromination Method):min. 98.0 %
- Melting point:109.0 to 113.0 deg-C
- Solubility in Water:almost transparency
Frequently Bought Together