
2-(2-Butoxyethoxy)ethyl acetate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:124-17-4
:
Formula:C₁₀H₂₀O₄
MDL Number:MFCD00009458
:204.27 g/mol
Boiling Pt:247°C
Melting Pt:–32°C
Density:0,973 g/cm³ (20 °C)
Flash Pt:124°C
EINECS:204-685-9
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together