
5-Hexen-2-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-49-9
:
Formula:C₆H₁₀O
MDL Number:MFCD00008793
:98.14 g/mol
Boiling Pt:129°C
Density:0,843 g/cm³ (20 °C)
Flash Pt:23°C
EINECS:203-675-1
UN:1224
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together