
Isopropyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4191-73-5
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00016468
:180.2 g/mol
Boiling Pt:160°C
EINECS:224-069-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to flake
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:84.0 to 87.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together