Order Entry
Italy
ContactUsLinkComponent
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
Irritant
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together