Order Entry
Italy
ContactUsLinkComponent
4,6-Dihydroxy-5-nitropyrimidine ≥98.0% (by titrimetric analysis)
Irritant
4,6-Dihydroxy-5-nitropyrimidine ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1960-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4,6-Dihydroxy-5-nitropyrimidine ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1960-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Amber powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :157.09 g/mol
Melting Pt:300 °C
EINECS:218-504-6
 :GHS07
CAS Number:2164-83-2
Formula:C₄H₃N₃O₄
MDL Number:MFCD00006110