
Isovaleric anhydride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1468-39-9
:
Formula:C₁₀H₁₈O₃
MDL Number:MFCD00015051
:186.25 g/mol
Boiling Pt:215°C
Melting Pt:–65 °C
Density:0,9327 g/cm³ (20 °C)
Flash Pt:91°C
EINECS:215-994-3
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
- Purity (Morpholine Method):min. 99.0 %
Specification Test Results
Frequently Bought Together