Order Entry
Italy
ContactUsLinkComponent
4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
Irritant
4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH0938-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAH0938-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    221.0 to 226.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :166.18 g/mol
 :GHS07
CAS Number:4919-37-3
Formula:C₉H₁₀O₃
MDL Number:MFCD00016536