3,3-Dimethylcyclohexanone ≥97.0% (by GC)
: TCI
- :2979-19-3
- :C₈H₁₄O
- :MFCD00040178
- :126.2 g/mol
- :175°C
- :6 °C
- :0,909 g/cm³ (25 °C)
- :59°C
- :1224
- :3,III
- :Density: 0.909 g/cm³ (25 °C)
- :2979-19-3
- :C₈H₁₄O
- :MFCD00040178
- :126.2 g/mol
- :175°C
- :6 °C
- :0,909 g/cm³ (25 °C)
- :59°C
- :1224
- :3,III
- :Density: 0.909 g/cm³ (25 °C)
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Product Family Options
- Pack typePk



