Order Entry
Italy
ContactUsLinkComponent
 
 
Thiophene-2-carboxylic hydrazide ≥98.0% (by HPLC)
  TCIAT1140-10G
 :  TCI
 :  
undefined
 
Thiophene-2-carboxylic hydrazide ≥98.0% (by HPLC)
  TCIAT1140-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    137.0 to 140.0 deg-C

 

 

 :
 :142.18 g/mol
 :142…143 °C
 :219-107-0
 :GHS07
 :2361-27-5
 :C₅H₆N₂OS
 :MFCD00005435